Product Details

SNP ID
rs150222211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180603181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGGAGATACCTAGTATGGATC[A/G]GAGAGAGCTGTTTTTCCGAGATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC14 PubMed Links

Gene Details

Gene
TTC14
Gene Name
tetratricopeptide repeat domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042601.2 476 Missense Mutation CAG,CGG Q115R NP_001036066.1
NM_001288582.1 476 Missense Mutation CAG,CGG Q115R NP_001275511.1
NM_133462.3 476 Missense Mutation CAG,CGG Q115R NP_597719.1
XM_011512439.2 476 Intron XP_011510741.1
XM_017005739.1 476 Missense Mutation CAG,CGG Q115R XP_016861228.1
XM_017005740.1 476 Missense Mutation CAG,CGG Q115R XP_016861229.1

View Full Product Details