Product Details

SNP ID
rs150260272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97943037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTTTAGTATTTCAATTTGAGT[C/T]ATAATAAGGTCCAATTTGAGGTGAA
Phenotype
MIM: 612049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYBG3 PubMed Links

Gene Details

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153605.3 3655 Intron NP_705833.3
XM_005247117.4 3655 Intron XP_005247174.1
Gene
MINA
Gene Name
MYC induced nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042533.2 3655 UTR 3 NP_001035998.1
NM_001261829.1 3655 UTR 3 NP_001248758.1
NM_032778.5 3655 UTR 3 NP_116167.3
NM_153182.3 3655 UTR 3 NP_694822.2
XM_005247838.4 3655 UTR 3 XP_005247895.1
XM_011513244.2 3655 Intron XP_011511546.1

View Full Product Details