Product Details

SNP ID
rs150323617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44594123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGACATTGCCCAGGAAGTCA[A/T]AATTTAGAAGGCTCCTCTGAAGGGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 263 UTR 5 NP_775929.2
XM_005265090.4 263 UTR 5 XP_005265147.1
XM_006713117.3 263 UTR 5 XP_006713180.1
XM_011533622.2 263 UTR 5 XP_011531924.1

View Full Product Details