Product Details

SNP ID
rs150551876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9893216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCTGCTGCACAGCAGCTCTAC[C/T]GCCATGGCAAGGCCTACCGTTTCCT
Phenotype
MIM: 614995 MIM: 616012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17RE PubMed Links

Gene Details

Gene
IL17RE
Gene Name
interleukin 17 receptor E
There are no transcripts associated with this gene.

Gene
JAGN1
Gene Name
jagunal homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032492.3 527 Missense Mutation CGC,TGC R131C NP_115881.3

View Full Product Details