Product Details

SNP ID
rs150743951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101652519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCATGGTTGTTCATGTGTCTC[C/T]TATACTCTCTTAGCTGAGTGAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB11 PubMed Links

Gene Details

Gene
ZBTB11
Gene Name
zinc finger and BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014415.3 4236 Missense Mutation AAG,AGG K874R NP_055230.2
XM_011512689.2 4236 Missense Mutation AAG,AGG K809R XP_011510991.1
XM_011512690.2 4236 Missense Mutation AAG,AGG K590R XP_011510992.1

View Full Product Details