Product Details

SNP ID
rs150810451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:16809735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGAATTGACATTGCATTGGCT[C/T]CTCTGACAGCCTTAAAGAGATACCA
Phenotype
MIM: 614276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCL2 PubMed Links

Gene Details

Gene
PLCL2
Gene Name
phospholipase C like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144382.1 Intron NP_001137854.1
NM_015184.5 Intron NP_055999.2
XM_006713073.3 Intron XP_006713136.1
XM_017006022.1 Intron XP_016861511.1
XM_017006023.1 Intron XP_016861512.1
XM_017006024.1 Intron XP_016861513.1
XM_017006025.1 Intron XP_016861514.1

View Full Product Details