Product Details

SNP ID
rs150852183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122806910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTATGGTTTTGGGAACTGGT[C/G]TAAGATGCATACCTATATCAGACTT
Phenotype
MIM: 602773
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIRC2 PubMed Links

Gene Details

Gene
DIRC2
Gene Name
disrupted in renal carcinoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032839.2 536 Missense Mutation CTA,GTA L133V NP_116228.1

View Full Product Details