Product Details

SNP ID
rs150870478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100368074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCAATAGTTCCCATGGTTTCA[C/T]AGGCAAAATCACATTTATTGTCAAT
Phenotype
MIM: 606081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM70 PubMed Links

Gene Details

Gene
TOMM70
Gene Name
translocase of outer mitochondrial membrane 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014820.4 2092 Missense Mutation TAT,TGT Y548C NP_055635.3

View Full Product Details