Product Details

SNP ID
rs150953894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151436743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCACTACCAAGTGGGTTCTTTGC[C/T]GTGCATTTGTATATCCCAGAATCGG
Phenotype
MIM: 611318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF10 PubMed Links

Gene Details

Gene
IGSF10
Gene Name
immunoglobulin superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178145.1 8627 Silent Mutation ACA,ACG T633T NP_001171616.1
NM_001178146.1 8627 Silent Mutation ACA,ACG T585T NP_001171617.1
NM_178822.4 8627 Silent Mutation ACA,ACG T2606T NP_849144.2
XM_011512708.2 8627 Silent Mutation ACA,ACG T2678T XP_011511010.1
XM_011512709.2 8627 Silent Mutation ACA,ACG T2606T XP_011511011.1
Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 8627 Intron NP_443728.3
XM_006713487.3 8627 Intron XP_006713550.1
XM_011512390.2 8627 Intron XP_011510692.1
XM_011512394.2 8627 Intron XP_011510696.1
XM_011512399.2 8627 Intron XP_011510701.1
XM_017005676.1 8627 Intron XP_016861165.1
XM_017005677.1 8627 Intron XP_016861166.1
XM_017005678.1 8627 Intron XP_016861167.1
XM_017005679.1 8627 Intron XP_016861168.1
XM_017005680.1 8627 Intron XP_016861169.1
XM_017005681.1 8627 Intron XP_016861170.1

View Full Product Details