Product Details

SNP ID
rs150994095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:107378030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACAGAACTGGAAATTCAGAATT[C/T]CTGCTCCACGATACATTGTCTAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC54 PubMed Links

Gene Details

Gene
CCDC54
Gene Name
coiled-coil domain containing 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032600.2 690 Missense Mutation TCC,TTC S148F NP_115989.1
Gene
LOC101929579
Gene Name
uncharacterized LOC101929579
There are no transcripts associated with this gene.

View Full Product Details