Product Details

SNP ID
rs151111656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185658353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCACTTACAGAGTGATGATG[C/T]GGGAACGGGCCAAACTGGTGATGGG
Phenotype
MIM: 608289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGF2BP2 PubMed Links

Gene Details

Gene
IGF2BP2
Gene Name
insulin like growth factor 2 mRNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007225.1 1361 Intron NP_001007226.1
NM_001291869.1 1361 Intron NP_001278798.1
NM_001291872.1 1361 Intron NP_001278801.1
NM_001291873.1 1361 Intron NP_001278802.1
NM_001291874.1 1361 Intron NP_001278803.1
NM_001291875.1 1361 Intron NP_001278804.1
NM_006548.4 1361 Intron NP_006539.3
XM_006713477.3 1361 Intron XP_006713540.1
XM_006713478.3 1361 Intron XP_006713541.1
XM_017005557.1 1361 Intron XP_016861046.1
XM_017005558.1 1361 Intron XP_016861047.1
XM_017005559.1 1361 Intron XP_016861048.1
XM_017005560.1 1361 Intron XP_016861049.1
XM_017005561.1 1361 Silent Mutation CCA,CCG P425P XP_016861050.1
XM_017005562.1 1361 Intron XP_016861051.1
XM_017005563.1 1361 Intron XP_016861052.1
XM_017005564.1 1361 Intron XP_016861053.1

View Full Product Details