Product Details

SNP ID
rs151156865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51944756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCCCTGCCCCGCTGCTCCTGCC[C/T]ACATGTGCCCTCTATCTTCAGGTGG
Phenotype
MIM: 607726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP3 PubMed Links

Gene Details

Gene
PARP3
Gene Name
poly(ADP-ribose) polymerase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003931.3 Intron NP_001003931.3
NM_005485.5 Intron NP_005476.4
XM_005264779.4 Intron XP_005264836.2
XM_017005490.1 Intron XP_016860979.1
Gene
RRP9
Gene Name
ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast)
There are no transcripts associated with this gene.

View Full Product Details