Product Details

SNP ID
rs151169020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48223595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCTGGTGATGCCTCTGGCCAT[C/T]ATTGCCCAGGTCCTCAGCTACAAGG
Phenotype
MIM: 600474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMP PubMed Links

Gene Details

Gene
CAMP
Gene Name
cathelicidin antimicrobial peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004345.4 249 Silent Mutation ATC,ATT I31I NP_004336.3

View Full Product Details