Product Details

SNP ID
rs151224158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44594558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCAGTGGAAAGTCACATCTTA[C/T]TCGGCACCAGGGAATCCACAGTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 698 Missense Mutation ACT,ATT T122I NP_775929.2
XM_005265090.4 698 Missense Mutation ACT,ATT T122I XP_005265147.1
XM_006713117.3 698 Missense Mutation ACT,ATT T122I XP_006713180.1
XM_011533622.2 698 Missense Mutation ACT,ATT T122I XP_011531924.1

View Full Product Details