Product Details

SNP ID
rs137905067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76895959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATCCAGTGCAACGCAGTGTACC[C/T]AGTCAAAAAGTCTTTGTGCAAGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100421494 PubMed Links

Gene Details

Gene
LOC100421494
Gene Name
microtubule affinity regulating kinase 2 pseudogene
There are no transcripts associated with this gene.

Gene
SOWAHB
Gene Name
sosondowah ankyrin repeat domain family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029870.1 1891 Missense Mutation AGG,GGG R631G NP_001025041.1

View Full Product Details