Product Details

SNP ID
rs137914743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174239060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGAATCTATGTGAGAACTTTG[A/T]TATAAGAGTTTCAAGATACTCTCCA
Phenotype
MIM: 605649
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO8 PubMed Links

Gene Details

Gene
FBXO8
Gene Name
F-box protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012180.2 1556 Missense Mutation ACA,TCA T236S NP_036312.2
XM_017008020.1 1556 Missense Mutation ACA,TCA T89S XP_016863509.1

View Full Product Details