Product Details

SNP ID
rs138067782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55951193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTTGTTTCCAGTTTGGGGCTAC[C/T]ATGAGTAAAGCTGTGAACATTCTTG
Phenotype
MIM: 611423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP135 PubMed Links

Gene Details

Gene
CEP135
Gene Name
centrosomal protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025009.4 Intron NP_079285.2
XM_005265788.3 Intron XP_005265845.1
XM_006714055.2 Intron XP_006714118.1
XM_011534412.2 Intron XP_011532714.1

View Full Product Details