Product Details
- SNP ID
-
rs138181423
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:24529675 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAAACTCATTATAAAGCACCCA[C/T]TCAGGTTTGTGGTCAAGAACAGTAG
- Phenotype
-
MIM: 603403
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DHX15
PubMed Links
Gene Details
- Gene
- DHX15
- Gene Name
- DEAH-box helicase 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001358.2 |
2364 |
Silent Mutation |
GAA,GAG |
E732E |
NP_001349.2 |
- Gene
- MIR573
- Gene Name
- microRNA 573
There are no transcripts associated with this gene.
View Full Product Details