Product Details

SNP ID
rs138191080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:147624884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGAAGCTTATGTAATTTACAA[C/T]TTTATGGGATTCCTTACCAATTATC
Phenotype
MIM: 613937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM184C PubMed Links

Gene Details

Gene
TMEM184C
Gene Name
transmembrane protein 184C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018241.2 941 Silent Mutation AAC,AAT N124N NP_060711.2

View Full Product Details