Product Details
- SNP ID
-
rs138440660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:78554487 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGGGATATCATGGCATCTATCT[C/G]GGTAAGTAAAAATTAAGCCTTCACA
- Phenotype
-
MIM: 106490
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA3
PubMed Links
Gene Details
- Gene
- ANXA3
- Gene Name
- annexin A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005139.2 |
324 |
Missense Mutation |
TCG,TGG |
S5W |
NP_005130.1 |
View Full Product Details