Product Details

SNP ID
rs138534367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139454330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGATCGGCGACTCCAATGTGGG[C/G]AAGACATGCCTGACCTACCGCTTCT
Phenotype
MIM: 605950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB33B PubMed Links

Gene Details

Gene
RAB33B
Gene Name
RAB33B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031296.2 620 Silent Mutation GGC,GGG G45G NP_112586.1
XM_011532299.1 620 Silent Mutation GGC,GGG G93G XP_011530601.1

View Full Product Details