Product Details

SNP ID
rs138727767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173333596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTCCCGGCAGGTCTGCACGAA[A/G]AAGGCGTACGAGGACATTTTGCCCC
Phenotype
MIM: 605005 MIM: 163906
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNT7 PubMed Links

Gene Details

Gene
GALNT7
Gene Name
polypeptide N-acetylgalactosaminyltransferase 7
There are no transcripts associated with this gene.

Gene
HMGB2
Gene Name
high mobility group box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130688.1 187 Silent Mutation TTC,TTT F18F NP_001124160.1
NM_001130689.1 187 Silent Mutation TTC,TTT F18F NP_001124161.1
NM_002129.3 187 Silent Mutation TTC,TTT F18F NP_002120.1
Gene
LOC105377538
Gene Name
uncharacterized LOC105377538
There are no transcripts associated with this gene.

View Full Product Details