Product Details

SNP ID
rs138871272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155928404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAAGAACTTCCCCAGGAATTC[C/T]GCACAATCCAATATGGAGTGCTTCC
Phenotype
MIM: 600550 MIM: 191070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSO PubMed Links

Gene Details

Gene
CTSO
Gene Name
cathepsin O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001334.2 912 Missense Mutation CAG,CGG Q288R NP_001325.1
Gene
TDO2
Gene Name
tryptophan 2,3-dioxygenase
There are no transcripts associated with this gene.

View Full Product Details