Product Details

SNP ID
rs139004992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75950767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAAGTTACTTCATTCTTTTTC[C/T]CTTTTTCAAAAGTGCATCTCGTAGT
Phenotype
MIM: 607469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAAA PubMed Links

Gene Details

Gene
NAAA
Gene Name
N-acylethanolamine acid amidase
There are no transcripts associated with this gene.

Gene
SDAD1
Gene Name
SDA1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288983.1 2224 Intron NP_001275912.1
NM_001288984.1 2224 Intron NP_001275913.1
NM_018115.3 2224 Intron NP_060585.2
XM_005263102.4 2224 Missense Mutation AGA,GGA R658G XP_005263159.1
XM_005263105.4 2224 Intron XP_005263162.1
XM_017008354.1 2224 Intron XP_016863843.1
XM_017008355.1 2224 Intron XP_016863844.1

View Full Product Details