Product Details

SNP ID
rs139181881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGGAGTACGTGGTGGTCGAGG[A/C]GGAGGAAGAGCTTACCCCCAGCAGT
Phenotype
MIM: 138297
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 860 Missense Mutation GAG,GCG E173A NP_001968.3
XM_017007877.1 860 Intron XP_016863366.1

View Full Product Details