Product Details

SNP ID
rs139310463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40425710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCTTTTGGAAATGTATGAACG[C/T]AGGCATGCTAAGTTGAAAATAGTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM47 PubMed Links

Gene Details

Gene
RBM47
Gene Name
RNA binding motif protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098634.1 2807 UTR 3 NP_001092104.1
NM_019027.3 2807 UTR 3 NP_061900.2
XM_005248103.3 2807 UTR 3 XP_005248160.1
XM_005248107.3 2807 UTR 3 XP_005248164.1
XM_005248108.3 2807 UTR 3 XP_005248165.1
XM_005248109.4 2807 UTR 3 XP_005248166.1
XM_011513704.2 2807 UTR 3 XP_011512006.1
XM_011513707.2 2807 UTR 3 XP_011512009.1
XM_011513708.2 2807 UTR 3 XP_011512010.1
XM_017008304.1 2807 UTR 3 XP_016863793.1
XM_017008305.1 2807 UTR 3 XP_016863794.1
XM_017008306.1 2807 UTR 3 XP_016863795.1
XM_017008307.1 2807 UTR 3 XP_016863796.1
XM_017008308.1 2807 UTR 3 XP_016863797.1
XM_017008309.1 2807 UTR 3 XP_016863798.1
XM_017008310.1 2807 UTR 3 XP_016863799.1

View Full Product Details