Product Details

SNP ID
rs139750456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2931128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGCTGCGCAGTGTACCCATGAC[C/T]GTGCCCTTCTGCCCTGGTGAGCCTG
Phenotype
MIM: 102680 MIM: 610977 MIM: 611526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADD1 PubMed Links

Gene Details

Gene
ADD1
Gene Name
adducin 1
There are no transcripts associated with this gene.

Gene
MFSD10
Gene Name
major facilitator superfamily domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001120.4 1292 Silent Mutation ACA,ACG T395T NP_001111.3
NM_001146069.1 1292 Silent Mutation ACA,ACG T395T NP_001139541.1
XM_005247927.3 1292 Silent Mutation ACA,ACG T395T XP_005247984.1
Gene
NOP14
Gene Name
NOP14 nucleolar protein
There are no transcripts associated with this gene.

Gene
NOP14-AS1
Gene Name
NOP14 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details