Product Details

SNP ID
rs139883264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:146640037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATACCATCCCGTGCACGTCGGC[C/T]GCCTCTTCTTCATCGGTGCCCATCT
Phenotype
MIM: 113725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POU4F2 PubMed Links

Gene Details

Gene
POU4F2
Gene Name
POU class 4 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004575.2 707 Silent Mutation GCC,GCT A153A NP_004566.2

View Full Product Details