Product Details

SNP ID
rs140157798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69844260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCATCTCTTGGAACGAAGTAT[A/G]ATGTATAATCCTGTCCACAAGCTCC
Phenotype
MIM: 600043
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT1E1 PubMed Links

Gene Details

Gene
SULT1E1
Gene Name
sulfotransferase family 1E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005420.2 786 Missense Mutation CAT,TAT H225Y NP_005411.1
XM_011532210.2 786 Intron XP_011530512.1

View Full Product Details