Product Details

SNP ID
rs140174752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89113655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTACACAAACTTAATCTTTGTGT[C/T]GTGGGGAAGGCCAAAAAGCCCCGAG
Phenotype
MIM: 613299 MIM: 612973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM13A PubMed Links

Gene Details

Gene
FAM13A
Gene Name
family with sequence similarity 13 member A
There are no transcripts associated with this gene.

Gene
TIGD2
Gene Name
tigger transposable element derived 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145715.2 839 Silent Mutation GTC,GTT V227V NP_663761.1

View Full Product Details