Product Details

SNP ID
rs140202402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154584197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATCTGCCCCTCTAAAGGAAACC[C/T]AGACCACTCCATTCTCAATCTCATA
Phenotype
MIM: 134820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGA PubMed Links

Gene Details

Gene
FGA
Gene Name
fibrinogen alpha chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000508.4 2607 Nonsense Mutation TAG,TGG *843W NP_000499.1
NM_021871.3 2607 Intron NP_068657.1

View Full Product Details