Product Details

SNP ID
rs140261968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:103082038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGATGAGCAAATTGTGGCCAAA[A/T]GTGATTGAGGGTAATGAACTCCTTG
Phenotype
MIM: 611789
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BDH2 PubMed Links

Gene Details

Gene
BDH2
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020139.3 Intron NP_064524.3
XM_005263140.3 Intron XP_005263197.1
XM_006714274.3 Intron XP_006714337.1
XM_011532127.2 Intron XP_011530429.1
XM_011532128.2 Intron XP_011530430.1
XM_017008461.1 Intron XP_016863950.1
XM_017008462.1 Intron XP_016863951.1
Gene
SLC9B2
Gene Name
solute carrier family 9 member B2
There are no transcripts associated with this gene.

View Full Product Details