Product Details

SNP ID
rs140327502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67619011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAACTTGTATTAATCAGTGTCAT[C/G]ACTGAAGTAGTATCTTACTGGAGGT
Phenotype
MIM: 611361
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UBA6 PubMed Links

Gene Details

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 3106 Missense Mutation CAT,GAT H1049D NP_060697.4
XM_017008359.1 3106 UTR 3 XP_016863848.1

View Full Product Details