Product Details

SNP ID
rs140417957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2937083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGCTCATGCTCTGGTCCAGC[C/T]ATGGCTTGAACACCTCCCAACAAGA
Phenotype
MIM: 102680 MIM: 610977 MIM: 611526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADD1 PubMed Links

Gene Details

Gene
ADD1
Gene Name
adducin 1
There are no transcripts associated with this gene.

Gene
MFSD10
Gene Name
major facilitator superfamily domain containing 10
There are no transcripts associated with this gene.

Gene
NOP14
Gene Name
NOP14 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291978.1 Intron NP_001278907.1
NM_001291979.1 Intron NP_001278908.1
NM_003703.2 Intron NP_003694.1
Gene
NOP14-AS1
Gene Name
NOP14 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details