Product Details
- SNP ID
-
rs140590165
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:155904087 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCACAAACTGGTGTGAATAGAGC[C/T]AGCAAAGGAGGTCTTATCTATGGGA
- Phenotype
-
MIM: 191070
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TDO2
PubMed Links
Gene Details
- Gene
- TDO2
- Gene Name
- tryptophan 2,3-dioxygenase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005651.3 |
171 |
Silent Mutation |
GCC,GCT |
A35A |
NP_005642.1 |
View Full Product Details