Product Details

SNP ID
rs140635234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37830078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACAGCTGGCCTCCGAGCTGCT[A/G]TGGGACCTGGAATTTCTCGTATGAA
Phenotype
MIM: 172000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGM2 PubMed Links

Gene Details

Gene
PGM2
Gene Name
phosphoglucomutase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018290.3 205 Missense Mutation ATG,GTG M66V NP_060760.2
XM_011513711.2 205 UTR 5 XP_011512013.1

View Full Product Details