Product Details

SNP ID
rs140803064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:80266858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTCCTCCTCCTCCTCTTCTTC[A/G]GCCACCTGATCCTCAGCGCCTGGGC
Phenotype
MIM: 165190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF5 PubMed Links

Gene Details

Gene
FGF5
Gene Name
fibroblast growth factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291812.1 271 Intron NP_001278741.1
NM_004464.3 271 Missense Mutation AGC,GGC S12G NP_004455.2
NM_033143.2 271 Missense Mutation AGC,GGC S12G NP_149134.1

View Full Product Details