Product Details
- SNP ID
-
rs140843867
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73076963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGTTTGAGGGCCATTATTTTCCG[G/T]GGAGCTGGAAACCATCTTTATCAGT
- Phenotype
-
MIM: 615929
MIM: 610428
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2306058] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD17
- Gene Name
- ankyrin repeat domain 17
- Gene
- COX18
- Gene Name
- COX18, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.
View Full Product Details