Product Details

SNP ID
rs141684609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185408850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGATGTCAAAGGTCTTCCTTGAC[C/T]ATCCAGTCTACAAACGGCCCACTCA
Phenotype
MIM: 611482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD37 PubMed Links

Gene Details

Gene
ANKRD37
Gene Name
ankyrin repeat domain 37
There are no transcripts associated with this gene.

Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114357.1 Intron NP_001107829.1
XM_005263019.3 Intron XP_005263076.1
XM_011531989.2 Intron XP_011530291.1
XM_017008236.1 Intron XP_016863725.1
XM_017008237.1 Intron XP_016863726.1
XM_017008238.1 Intron XP_016863727.1
XM_017008239.1 Intron XP_016863728.1
XM_017008240.1 Intron XP_016863729.1
XM_017008241.1 Intron XP_016863730.1
XM_017008242.1 Intron XP_016863731.1
Gene
UFSP2
Gene Name
UFM1 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018359.3 Intron NP_060829.2

View Full Product Details