Product Details
- SNP ID
-
rs141947839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:849961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTTGCTCCGGAGCCACCAGGTC[A/G]GCCATGCCCACGGGCGTCCAGCGGC
- Phenotype
-
MIM: 602052
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAK
PubMed Links
Gene Details
- Gene
- GAK
- Gene Name
- cyclin G associated kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318134.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1176A |
NP_001305063.1 |
NM_005255.3 |
4040 |
Silent Mutation |
GCC,GCT |
A1255A |
NP_005246.2 |
XM_005272268.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1244A |
XP_005272325.1 |
XM_005272270.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1180A |
XP_005272327.1 |
XM_011513425.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1299A |
XP_011511727.1 |
XM_011513426.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1288A |
XP_011511728.1 |
XM_011513427.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1269A |
XP_011511729.1 |
XM_011513428.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1250A |
XP_011511730.1 |
XM_011513429.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1237A |
XP_011511731.1 |
XM_011513430.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1220A |
XP_011511732.1 |
XM_011513431.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1216A |
XP_011511733.1 |
XM_011513432.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1211A |
XP_011511734.1 |
XM_011513434.2 |
4040 |
Silent Mutation |
GCC,GCT |
A1163A |
XP_011511736.1 |
XM_017007991.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1225A |
XP_016863480.1 |
XM_017007992.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1190A |
XP_016863481.1 |
XM_017007993.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1163A |
XP_016863482.1 |
XM_017007994.1 |
4040 |
Intron |
|
|
XP_016863483.1 |
XM_017007995.1 |
4040 |
Silent Mutation |
GCC,GCT |
A1075A |
XP_016863484.1 |
View Full Product Details