Product Details

SNP ID
rs142193683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:117085075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCTTTGGTGAACTGCATTCTCT[G/T]GTTAATTTTATCCAACACATATTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAM1L1 PubMed Links

Gene Details

Gene
TRAM1L1
Gene Name
translocation associated membrane protein 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152402.2 506 Missense Mutation AAG,CAG K107Q NP_689615.2

View Full Product Details