Product Details
- SNP ID
-
rs142276783
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:105897987 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGGGGCTGGGCTCGCCAGTCTT[C/G]GGGACAGTGTCAGCGTGAGTATCAA
- Phenotype
-
MIM: 610306
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPNT
PubMed Links
Gene Details
- Gene
- NPNT
- Gene Name
- nephronectin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033047.2 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
NP_001028219.1 |
NM_001184690.1 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
NP_001171619.1 |
NM_001184691.1 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
NP_001171620.1 |
NM_001184692.1 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
NP_001171621.1 |
NM_001184693.1 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
NP_001171622.1 |
XM_005262888.4 |
371 |
Missense Mutation |
TCG,TGG |
S53W |
XP_005262945.1 |
XM_005262890.1 |
371 |
Intron |
|
|
XP_005262947.1 |
XM_011531820.2 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_011530122.1 |
XM_011531822.2 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_011530124.1 |
XM_011531823.2 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_011530125.1 |
XM_011531824.2 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_011530126.1 |
XM_011531825.2 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_011530127.1 |
XM_017007984.1 |
371 |
Missense Mutation |
TCG,TGG |
S54W |
XP_016863473.1 |
View Full Product Details