Product Details

SNP ID
rs142543281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39499665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAAATTCTAGATTCATAATTA[C/T]CAGATATGCTAATATCCATATAGCT
Phenotype
MIM: 603370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGDH PubMed Links

Gene Details

Gene
UGDH
Gene Name
UDP-glucose 6-dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184700.1 2286 UTR 3 NP_001171629.1
NM_001184701.1 2286 UTR 3 NP_001171630.1
NM_003359.3 2286 UTR 3 NP_003350.1
XM_005262667.2 2286 UTR 3 XP_005262724.1

View Full Product Details