Product Details

SNP ID
rs142803417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5898717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAATACTGACTCAAAGTTAGT[A/G]TCAAGTTTTCAGCCATTTTTAACAA
Phenotype
MIM: 602462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf50 PubMed Links

Gene Details

Gene
C4orf50
Gene Name
chromosome 4 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011513603.2 13348 UTR 3 XP_011511905.1
XM_011513604.2 13348 UTR 3 XP_011511906.1
XM_017008891.1 13348 UTR 3 XP_016864380.1
XM_017008892.1 13348 UTR 3 XP_016864381.1
XM_017008893.1 13348 UTR 3 XP_016864382.1
XM_017008894.1 13348 Intron XP_016864383.1
Gene
CRMP1
Gene Name
collapsin response mediator protein 1
There are no transcripts associated with this gene.

View Full Product Details