Product Details

SNP ID
rs142989025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152769634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCCTGGAGATCCATCATCATC[C/G]TCTTCATCAGAAGTGTAAAATCCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TIGD4 PubMed Links

Gene Details

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 2192 Missense Mutation GAC,GAG D457E NP_663772.1
XM_005262807.4 2192 Missense Mutation GAC,GAG D457E XP_005262864.1

View Full Product Details