Product Details

SNP ID
rs143107150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTATCTCCTGACCATGGAGTTC[C/G]CCGGCTGGCTGAACGGCTCCCTCGT
Phenotype
MIM: 138297
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 919 Missense Mutation CCC,GCC P193A NP_001968.3
XM_017007877.1 919 Intron XP_016863366.1

View Full Product Details