Product Details

SNP ID
rs143129444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70637839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACCAATCAGCAATGAAGAAG[G/T]GGGGGTAAGTACAAGTAAAACTACA
Phenotype
MIM: 606585
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ENAM PubMed Links

Gene Details

Gene
ENAM
Gene Name
enamelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031889.2 2614 Missense Mutation GGG,GTG G195V NP_114095.2
XM_006714056.3 2614 Missense Mutation GGG,GTG G195V XP_006714119.1

View Full Product Details