Product Details

SNP ID
rs143207059
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99549153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTATCCTGCTTTTCTTCATGTG[G/T]TGAATCTAGTTCATTTCTGCTATAT
Phenotype
MIM: 616013
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C4orf17 PubMed Links

Gene Details

Gene
C4orf17
Gene Name
chromosome 4 open reading frame 17
There are no transcripts associated with this gene.

Gene
TRMT10A
Gene Name
tRNA methyltransferase 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134665.2 1102 Missense Mutation ACA,CCA T319P NP_001128137.1
NM_001134666.2 1102 Missense Mutation ACA,CCA T319P NP_001128138.1
NM_152292.4 1102 Missense Mutation ACA,CCA T319P NP_689505.1
XM_005263352.3 1102 Missense Mutation ACA,CCA T319P XP_005263409.1
XM_006714417.2 1102 Missense Mutation ACA,CCA T319P XP_006714480.1

View Full Product Details