Product Details

SNP ID
rs143222398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:147624991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCCCTCCTTTATGTTGCTGTC[C/T]ACCATGGGCTATGGGAGAGTAAGTA
Phenotype
MIM: 613937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM184C PubMed Links

Gene Details

Gene
TMEM184C
Gene Name
transmembrane protein 184C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018241.2 1048 Missense Mutation CCA,CTA P160L NP_060711.2

View Full Product Details