Product Details

SNP ID
rs143338080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68229263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTACAAAAAAACTTACCATTCA[C/T]ATAAAGTGTTCCCCAACCTGTAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11B PubMed Links

Gene Details

Gene
TMPRSS11B
Gene Name
transmembrane protease, serine 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182502.3 912 Missense Mutation ATG,GTG M314V NP_872308.2
XM_011531608.2 912 Missense Mutation ATG,GTG M314V XP_011529910.1
XM_011531609.1 912 UTR 3 XP_011529911.1

View Full Product Details